Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-5-nitrothiophene-3-carboxylate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-5-nitrothiophene-3-carboxylate
CAS 42783-04-0
Formula Weight 216.21
MDL Number MFCD01566377
Synonym 2-Amino-5-nitrothiophene-3-carboxylic acid ethyl ester; 2-Amino-3-ethoxycarbonyl-5-nitrothiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6162503

  SDS
alfa aesar™
H6162503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61625MD

  SDS
alfa aesar™
H61625MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.21
Quantity 1g
MDL Number MFCD01566377
Chemical Name or Material Ethyl 2-amino-5-nitrothiophene-3-carboxylate
Assay 95%
Synonym 2-Amino-5-nitrothiophene-3-carboxylic acid ethyl ester; 2-Amino-3-ethoxycarbonyl-5-nitrothiophene
Physical Form Solid
CAS 42783-04-0