missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-amino-5-nitrothiophene-3-carboxylate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6162503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-amino-5-nitrothiophene-3-carboxylate | |
C7H8N2O4S | |
1 g | |
UXDKVBJDZDRIKL-UHFFFAOYSA-N | |
ethyl 2-amino-5-nitrothiophene-3-carboxylate | |
2737608 | |
95% |
42783-04-0 | |
MFCD01566377 | |
2-amino-3-ethoxycarbonyl-5-nitrothiophene, 3-thiophenecarboxylicacid, 2-amino-5-nitro-, ethyl ester, 2-amino-3-carbethoxy-5-nitrothiophene;, 2-amino-3-carboethoxy-5-nitrothiophene;, 2-amino-3-ethoxycarbonyl-5-nitrothiophene;, 2-amino-5-nitro-3-ethoxycarbonyl-thiophene, 2-amino-5-nitrothiophen-3-carbonsaeureethylester, 2-amino-5-nitro-3-thiophenecarboxylic acid ethyl ester, 2-amino-5-nitro-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid,2-amino-5-nitro-,ethyl ester | |
CCOC(=O)C1=C(SC(=C1)[N+](=O)[O-])N | |
216.211 | |
216.21 | |
Solid |
Chemical Identifiers
42783-04-0 | |
216.211 | |
UXDKVBJDZDRIKL-UHFFFAOYSA-N | |
2737608 | |
CCOC(=O)C1=C(SC(=C1)[N+](=O)[O-])N |
C7H8N2O4S | |
MFCD01566377 | |
2-amino-3-ethoxycarbonyl-5-nitrothiophene, 3-thiophenecarboxylicacid, 2-amino-5-nitro-, ethyl ester, 2-amino-3-carbethoxy-5-nitrothiophene;, 2-amino-3-carboethoxy-5-nitrothiophene;, 2-amino-3-ethoxycarbonyl-5-nitrothiophene;, 2-amino-5-nitro-3-ethoxycarbonyl-thiophene, 2-amino-5-nitrothiophen-3-carbonsaeureethylester, 2-amino-5-nitro-3-thiophenecarboxylic acid ethyl ester, 2-amino-5-nitro-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid,2-amino-5-nitro-,ethyl ester | |
ethyl 2-amino-5-nitrothiophene-3-carboxylate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 610-058-4
TSCA : No
Recommended Storage : Ambient temperatures