missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Amino-5-methylthiophene-3-carboxylate, TCI America™

Manufacturer:  TCI America E11045G

 View more versions of this product

Catalog No. E11045G



Specifications

Specifications

Ethyl 2-Amino-5-methylthiophene-3-carboxylate
49°C
4815-32-1
MFCD01922142
AYBUNZGJQVYGTM-UHFFFAOYSA-N
ethyl 2-amino-5-methylthiophene-3-carboxylate
4062885
Crystal-Powder at 20°C
Yellow
5g
C8H11NO2S
2-amino-5-methyl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-, ethyl ester, ethyl2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methyl-3-thiophenecarboxylic acid ethyl ester, ethyl 2-amino-5-methyl-thiophene-3-carboxylate, ksc590m8t, aronis25101, 2-amino-5-methyl-thiophene-3-carboxylicacidethylester
CCOC(=O)C1=C(SC(=C1)C)N
185.241
185.24
≥98.0% (GC)

Chemical Identifiers

4815-32-1
185.241
AYBUNZGJQVYGTM-UHFFFAOYSA-N
4062885
CCOC(=O)C1=C(SC(=C1)C)N
C8H11NO2S
MFCD01922142
2-amino-5-methyl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-, ethyl ester, ethyl2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methyl-3-thiophenecarboxylic acid ethyl ester, ethyl 2-amino-5-methyl-thiophene-3-carboxylate, ksc590m8t, aronis25101, 2-amino-5-methyl-thiophene-3-carboxylicacidethylester
ethyl 2-amino-5-methylthiophene-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents