Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-5-methylthiophene-3-carboxylate
CAS 4815-32-1
Formula Weight 185.24
MDL Number MFCD01922142
Synonym 2-Amino-5-methylthiophene-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6176603

 
alfa aesar™
H6176603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6176606

 
alfa aesar™
H6176606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.24
Physical Form Solid
Chemical Name or Material Ethyl 2-amino-5-methylthiophene-3-carboxylate
Synonym 2-Amino-5-methylthiophene-3-carboxylic acid ethyl ester
Assay 95%
MDL Number MFCD01922142
CAS 4815-32-1
Quantity 1g