Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%

$93.27 - $93.27

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
CAS 4815-37-6
Molecular Formula C14H15NO2S
Formula Weight 261.34
MDL Number MFCD01763475
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5179903

  SDS
alfa aesar™
H5179903
1g Each for $93.27
Description & Specifications

Specifications

Formula Weight 261.34
CAS 4815-37-6
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C14H15NO2S
Chemical Name or Material Ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Assay 97%
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
MDL Number MFCD01763475
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.