Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%

$83.93 - $1,279.24

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
CAS 149771-09-5
Molecular Formula C8H8F3N3O2
Formula Weight 235.17
MDL Number MFCD00052070
Synonym 2-Amino-4-(trifluoromethyl)pyrimidine-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2072403

  SDS
alfa aesar™
B2072403
1g Each for $83.93

AAB2072406

  SDS
alfa aesar™
B2072406
5g Each for $308.89

AAB2072414

  SDS
alfa aesar™
B2072414
25g Each for $1,279.24
Description & Specifications

Specifications

Formula Weight 235.17
Synonym 2-Amino-4-(trifluoromethyl)pyrimidine-5-carboxylic acid ethyl ester
CAS 149771-09-5
Molecular Formula C8H8F3N3O2
Chemical Name or Material Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
MDL Number MFCD00052070
Quantity 1g
Melting Point 179° to 180°C
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.