Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-4-phenylthiazole-5-carboxylate, 98%

$63.14 - $207.93

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-4-phenylthiazole-5-carboxylate
CAS 64399-23-1
Molecular Formula C12H12N2O2S
Formula Weight 248.31
MDL Number MFCD00022452
Beilstein 222259
Synonym 2-Amino-4-phenylthiazole-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1386403

  SDS
alfa aesar™
L1386403
1g Each for $63.14

AAL1386406

  SDS
alfa aesar™
L1386406
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 248.31
Chemical Name or Material Ethyl 2-amino-4-phenylthiazole-5-carboxylate
Assay 98%
MDL Number MFCD00022452
CAS 64399-23-1
Synonym 2-Amino-4-phenylthiazole-5-carboxylic acid ethyl ester
Molecular Formula C12H12N2O2S
Beilstein 222259
Melting Point 170° to 173°C
Quantity 1g