Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-4,5-diphenylthiophene-3-carboxylate, 97%

$362.91 - $362.91

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-4,5-diphenylthiophene-3-carboxylate
CAS 4815-43-4
Molecular Formula C19H17NO2S
Formula Weight 323.41g/mol
MDL Number MFCD00688367
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5184606

 
alfa aesar™
H51846-06
Each for $362.91
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Ethyl 2-amino-4,5-diphenylthiophene-3-carboxylate
CAS 4815-43-4
Assay 97%
Molecular Formula C19H17NO2S
Formula Weight 323.41g/mol
MDL Number MFCD00688367
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.