Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amino-4,5-dimethoxybenzoate, 98%

$67.11 - $832.76

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-4,5-dimethoxybenzoate
CAS 20323-74-4
Molecular Formula C11H15NO4
Formula Weight 225.24
MDL Number MFCD00017323
Beilstein 2646663
Synonym 2-Amino-4,5-dimethoxybenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0245703

 
alfa aesar™
L0245703
1g Each for $67.11

AAL0245706

 
alfa aesar™
L0245706
5g Each for $218.58

AAL0245714

 
alfa aesar™
L0245714
25g Each for $832.76
Description & Specifications

Specifications

Formula Weight 225.24
Chemical Name or Material Ethyl 2-amino-4,5-dimethoxybenzoate
Assay 98%
Molecular Formula C11H15NO4
MDL Number MFCD00017323
CAS 20323-74-4
Beilstein 2646663
Synonym 2-Amino-4,5-dimethoxybenzoic acid ethyl ester
Melting Point 86° to 88°C
Quantity 1g