Please login to your online account to display your discounted pricing

Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate
Name Note 99%
CAS 4506-71-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H15NO2S
Formula Weight 225.31
Synonym 2-Amino-3-carbethoxy-4, 5-tetramethylenethiophene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC251280010

  SDS
thermo fisher scientific
AC251280010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC251280100

  SDS
thermo fisher scientific
AC251280100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 225.31
Quantity 1g
Color Yellow
EINECS Number 224-823-1
Melting Point 114° to 117°C
Chemical Name or Material Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H15NO2S
CAS 4506-71-2
Synonym 2-Amino-3-carbethoxy-4, 5-tetramethylenethiophene
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.