missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%, ACROS Organics™

$75.79 - $75.79

Chemical Identifiers

CAS 7149-18-0
Assay Percent Range 99%
Molecular Formula C8H13NO2
Formula Weight 155.2
MDL Number MFCD00216929
Chemical Name or Material Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC292775000 SDS Thermo Fisher Scientific
AC292775000
Glass bottle 500mg Each for $75.79
Specifications Description & Specifications

Chemical Identifiers

7149-18-0
C8H13NO2
MFCD00216929
≥98.5%
99%
155.2
Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%

Specifications

Crystalline Powder
500mg
57° to 60°C
Glass bottle
7149-18-0
MFCD00216929
≥98.5%
155.2
Light yellow
Authentic
C8H13NO2
99%
Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.