Learn More
Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 292775000
Specifications
Ethyl 2-amino-1-cyclopentene-1-carboxylate | |
7149-18-0 | |
Yellow | |
98.5% min. (GC) | |
C8H13NO2 | |
500 mg | |
Solubility in water: soluble. Other solubilities: soluble in chloroform,methanol and ethanol | |
CCOC(=O)C1=C(N)CCC1 | |
155.20 | |
155.2 | |
Crystalline Powder |
99% | |
57°C to 60°C | |
Authentic | |
Glass bottle | |
MFCD00216929 | |
ethyl 2-amino-1-cyclopentene-1-carboxylate, ethyl 2-aminocyclopent-1-enecarboxylate, 2-amino-cyclopent-1-enecarboxylic acid ethyl ester, ethyl 2-aminocyclopent-1-ene-1-carboxylate, 2-amino-cyclopent-1-enecarboxylicacidethylester, ethyl trans-2-aminocyclopentenecarboxylate, ethyl 2-amino-1-cyclopentene-1-carboxylate #, 2-aminocyclopentene-1-carboxylic acid ethyl ester, cyclopentene-1-carboxylic acid, 2-amino-, ethyl ester, 1-cyclopentene-1-carboxylicacid, 2-amino-, ethyl ester | |
FMEHILPGLKGSCA-UHFFFAOYSA-N | |
ethyl 2-aminocyclopentene-1-carboxylate | |
243333 | |
99% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only