Please login to your online account to display your discounted pricing

Ethyl 2-amino-1-cyclohexene-1-carboxylate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-amino-1-cyclohexene-1-carboxylate
Name Note 98%
CAS 1128-00-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H15NO2
Linear Formula NHC6H8COOCHCH3
Formula Weight 169.22
MDL Number MFCD01863243
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334260010

  SDS
fisher scientific acros
334260010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 169.22
Quantity 1g
Color White to light yellow
Melting Point 70° to 74°C
Chemical Name or Material Ethyl 2-amino-1-cyclohexene-1-carboxylate
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H15NO2
Linear Formula NHC6H8COOCHCH3
MDL Number MFCD01863243
CAS 1128-00-3
Health Hazard 1 LIGHT SENSITIVE
Infrared Spectrum Authentic
Safety and Handling

LIGHT SENSITIVE