missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-amidinoacetate hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H6428803

 View more versions of this product

Catalog No. AAH6428803



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Ethyl 2-amidinoacetate hydrochloride
1g
C5H11ClN2O2
3694030
VOHFLYOSVGWQOS-UHFFFAOYSA-N
ethyl 3-amino-3-iminopropanoate;hydrochloride
15555354
Solid
105°C to 108°C
57508-48-2
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
CCOC(=O)CC(=N)N.Cl
166.605
166.6
95%

Chemical Identifiers

57508-48-2
166.605
VOHFLYOSVGWQOS-UHFFFAOYSA-N
15555354
CCOC(=O)CC(=N)N.Cl
C5H11ClN2O2
MFCD06797615
ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
ethyl 3-amino-3-iminopropanoate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents