missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Acetyl-3-ethoxyacrylate, TCI America™

Manufacturer:  TCI America E114825G

 View more versions of this product

Catalog No. E114825G



Specifications

Specifications

Ethyl 2-Acetyl-3-ethoxyacrylate
Undesignated
3788-94-1
MFCD06797311
FNASCUBBFNCFQO-VURMDHGXSA-N
ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate
5355337
Clear Liquid at 20°C
266°C
25g
C9H14O4
2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester
CCOC=C(C(=O)C)C(=O)OCC
186.207
186.21
≥98.0% (GC)

Chemical Identifiers

3788-94-1
186.207
FNASCUBBFNCFQO-VURMDHGXSA-N
5355337
CCOC=C(C(=O)C)C(=O)OCC
C9H14O4
MFCD06797311
2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester
ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents