missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Acetyl-3-(dimethylamino)acrylate, TCI America™

Manufacturer:  TCI America E117925G

 View more versions of this product

Catalog No. E117925G



Specifications

Specifications

Ethyl 2-Acetyl-3-(dimethylamino)acrylate
Undesignated
51145-57-4
MFCD00087563
LQSOVGAUOHMPLK-SOFGYWHQSA-N
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
2763877
Clear Liquid at 20°C
150°C
25g
C9H15NO3
ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
CCOC(=O)C(=CN(C)C)C(=O)C
185.223
185.22
≥95.0% (GC,T)

Chemical Identifiers

51145-57-4
185.223
LQSOVGAUOHMPLK-SOFGYWHQSA-N
2763877
CCOC(=O)C(=CN(C)C)C(=O)C
C9H15NO3
MFCD00087563
ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents