missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%

Manufacturer:  Alfa Aesar™ H6407303

 View more versions of this product

Catalog No. AAH6407303



Specifications

Specifications

Ethyl 2-acetyl-3-(dimethylamino)acrylate
1g
C9H15NO3
ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
CCOC(=O)C(=CN(C)C)C(=O)C
185.223
185.22
95%
Red
51145-57-4
MFCD00087563
LQSOVGAUOHMPLK-SOFGYWHQSA-N
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
2763877
Liquid

Chemical Identifiers

51145-57-4
185.223
LQSOVGAUOHMPLK-SOFGYWHQSA-N
2763877
CCOC(=O)C(=CN(C)C)C(=O)C
C9H15NO3
MFCD00087563
ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
SDS Documents