missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6407306

 View more versions of this product

Catalog No. AAH6407306



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 2-acetyl-3-(dimethylamino)acrylate
Red
MFCD00087563
ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
CCOC(=O)C(=CN(C)C)C(=O)C
185.223
185.22
Liquid
51145-57-4
C9H15NO3
5 g
LQSOVGAUOHMPLK-SOFGYWHQSA-N
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
2763877
95%

Chemical Identifiers

51145-57-4
185.223
LQSOVGAUOHMPLK-SOFGYWHQSA-N
ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
C9H15NO3
MFCD00087563
2763877
CCOC(=O)C(=CN(C)C)C(=O)C
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only