Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,6-dimethylbenzoate, 98%

$79.58 - $267.84

Chemical Identifiers

Chemical Name or Material Ethyl 2,6-dimethylbenzoate
CAS 36596-67-5
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00051785
Beilstein 2575407
Synonym 2,6-Dimethylbenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0738103

 
alfa aesar™
L0738103
1g Each for $79.58

AAL0738106

 
alfa aesar™
L0738106
5g Each for $267.84
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Ethyl 2,6-dimethylbenzoate
Boiling Point 78° to 80°C (0.9mmHg)
Assay 98%
Molecular Formula C11H14O2
MDL Number MFCD00051785
CAS 36596-67-5
Quantity 1g
Beilstein 2575407
Synonym 2,6-Dimethylbenzoic acid ethyl ester

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.