Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (2,6-dimethoxyphenoxy)acetate

$205.27 - $205.27

Chemical Identifiers

Chemical Name or Material Ethyl (2,6-dimethoxyphenoxy)acetate
CAS 15267-83-1
Molecular Formula C12H16O5
Formula Weight 240.26g/mol
MDL Number MFCD08064243
Synonym 2,6-Dimethoxyphenoxyacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5075203

 
alfa aesar™
H50752-03
1g Each for $205.27
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material Ethyl (2,6-dimethoxyphenoxy)acetate
CAS 15267-83-1
Molecular Formula C12H16O5
Formula Weight 240.26g/mol
MDL Number MFCD08064243
Synonym 2,6-Dimethoxyphenoxyacetic acid ethyl ester