Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,6-dichlorobenzoate, 97%

$67.58 - $862.00

Chemical Identifiers

Chemical Name or Material Ethyl 2,6-dichlorobenzoate
CAS 81055-73-4
Molecular Formula C9H8Cl2O2
Formula Weight 219.07
MDL Number MFCD01313702
Synonym 2,6-Dichlorobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2527503

  SDS
alfa aesar™
B2527503
1g Each for $67.58

AAB2527506

  SDS
alfa aesar™
B2527506
5g Each for $240.12

AAB2527514

  SDS
alfa aesar™
B2527514
25g Each for $862.00
Description & Specifications

Specifications

Formula Weight 219.07
Chemical Name or Material Ethyl 2,6-dichlorobenzoate
Assay 97%
Molecular Formula C9H8Cl2O2
MDL Number MFCD01313702
CAS 81055-73-4
Synonym 2,6-Dichlorobenzoic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.