missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%

Manufacturer:  Alfa Aesar™ H54192MD

 View more versions of this product

Catalog No. AAH54192MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 2-(4-pyridyl)thiazole-4-acetate
80653-68-5
MFCD06739821
JZBJUBBYJUUKQK-UHFFFAOYSA-N
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
7176350
97%
250mg
C12H12N2O2S
2-4-pyridyl thiazole-4-acetic acid ethyl ester
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2
248.3
248.3

Chemical Identifiers

80653-68-5
248.3
JZBJUBBYJUUKQK-UHFFFAOYSA-N
7176350
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2
C12H12N2O2S
MFCD06739821
2-4-pyridyl thiazole-4-acetic acid ethyl ester
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents