missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%

Manufacturer:  Alfa Aesar™ H5419203

 View more versions of this product

Catalog No. AAH5419203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 2-(4-pyridyl)thiazole-4-acetate
80653-68-5
MFCD06739821
JZBJUBBYJUUKQK-UHFFFAOYSA-N
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
7176350
97%
1g
C12H12N2O2S
2-4-pyridyl thiazole-4-acetic acid ethyl ester
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2
248.3
248.3

Chemical Identifiers

80653-68-5
248.3
JZBJUBBYJUUKQK-UHFFFAOYSA-N
7176350
CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2
C12H12N2O2S
MFCD06739821
2-4-pyridyl thiazole-4-acetic acid ethyl ester
ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents