missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-(4-methylperhydro-1,4-diazepin-1-yl)benzoate, 97%, Maybridge

Chemical Identifiers

CAS 912569-58-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H22N2O2
Formula Weight 262.35
MDL Number MFCD09025904
Chemical Name or Material Ethyl 2-(4-methylperhydro-1, 4-diazepin-1-yl)benzoate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60423CB SDS Fisher Scientific Company
CC60423CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60423DA SDS Fisher Scientific Company
CC60423DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

912569-58-5
100.0
C15H22N2O2
MFCD09025904
97%
96.5
97%
262.35
Ethyl 2-(4-methylperhydro-1, 4-diazepin-1-yl)benzoate

Specifications

250mg
96.5
Amber Glass Bottle
C15H22N2O2
MFCD09025904
97%
912569-58-5
100.0
97%
262.35
Ethyl 2-(4-methylperhydro-1, 4-diazepin-1-yl)benzoate

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger