missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4-dioxoheptanoate, 95%

$37.70 - $149.59

Chemical Identifiers

CAS 36983-31-0
Molecular Formula C9H14O4
Formula Weight 186.21
MDL Number MFCD08700411
Synonym 2,4-Dioxoheptanoic acid ethyl ester
Chemical Name or Material Ethyl 2,4-dioxoheptanoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3102703 SDS Alfa Aesar™
H3102703
1g Each for $37.70
AAH3102706 SDS Alfa Aesar™
H3102706
5g Each for $149.59
AAH3102714 SDS Alfa Aesar™
H3102714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

36983-31-0
186.21
2,4-Dioxoheptanoic acid ethyl ester
C9H14O4
MFCD08700411
Ethyl 2,4-dioxoheptanoate

Specifications

1g
228° to 232°C
186.21
2,4-Dioxoheptanoic acid ethyl ester
95%
36983-31-0
C9H14O4
MFCD08700411
Ethyl 2,4-dioxoheptanoate