Please login to your online account to display your discounted pricing

Ethyl 2,4-dioxo-4-(2-thienyl)butanoate, 90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB04027DA

 View more versions of this product

Catalog No. SPB04027DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.25
Assay 90%
Quantity 1g
CAS Min % 90.0
CAS 36983-36-5
Assay Percent Range 90%
MDL Number MFCD00126316
Chemical Name or Material Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate
Packaging Amber Glass Bottle
Molecular Formula C10H10O4S

Specifications

Chemical Name or Material Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate
CAS 36983-36-5
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C10H10O4S
Formula Weight 226.25
MDL Number MFCD00126316