Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4-dimethylthiazole-5-carboxylate, 97%

$347.88 - $347.88

Chemical Identifiers

Chemical Name or Material Ethyl 2,4-dimethylthiazole-5-carboxylate
CAS 7210-77-7
Molecular Formula C8H11NO2S
Formula Weight 185.24
MDL Number MFCD00052874
Synonym 2,4-Dimethylthiazole-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2086409

  SDS
alfa aesar™
B2086409
10g Each for $347.88
Description & Specifications

Specifications

Formula Weight 185.24
Chemical Name or Material Ethyl 2,4-dimethylthiazole-5-carboxylate
Assay 97%
Molecular Formula C8H11NO2S
MDL Number MFCD00052874
CAS 7210-77-7
Synonym 2,4-Dimethylthiazole-5-carboxylic acid ethyl ester
Melting Point 48° to 50°C
Quantity 10g