Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4-dimethylnicotinate, 96%

$88.15 - $299.85

Chemical Identifiers

Chemical Name or Material Ethyl 2,4-dimethylnicotinate
CAS 37669-78-6
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00051962
Beilstein 133910
Synonym Ethyl 2,4-dimethylpyridine-3-carboxylate; 2,4-Dimethylpyridine-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0867203

  SDS
alfa aesar™
L0867203
1g Each for $88.15

AAL0867206

  SDS
alfa aesar™
L0867206
5g Each for $299.85
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material Ethyl 2,4-dimethylnicotinate
Assay 96%
MDL Number MFCD00051962
CAS 37669-78-6
Molecular Formula C10H13NO2
Beilstein 133910
Synonym Ethyl 2,4-dimethylpyridine-3-carboxylate; 2,4-Dimethylpyridine-3-carboxylic acid ethyl ester
Boiling Point 116° to 118°C (10mmHg)
Quantity 1g

Applications
It is employed in the design, synthesis, and evaluation of a family of propargyl pyridinyl ethers as potential cytochrome p450 inhibitors.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.