missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2,4-dimethylbenzoate, 98+%, Thermo Scientific™
$89.11 - $302.44
Chemical Identifiers
CAS | 33499-42-2 |
---|---|
Molecular Formula | C11H14O2 |
Molecular Weight (g/mol) | 178.231 |
MDL Number | MFCD00015438 |
InChI Key | RJYPOWRKMKNFHH-UHFFFAOYSA-N |
Synonym | 2,4-dimethylbenzoic acid ethyl ester, benzoic acid, 2,4-dimethyl-, ethyl ester, acmc-20al1j, 2,4-dimethyl-benzoic acid ethyl ester, benzoic acid,2,4-dimethyl-, ethyl ester |
PubChem CID | 118492 |
IUPAC Name | ethyl 2,4-dimethylbenzoate |
SMILES | CCOC(=O)C1=C(C=C(C=C1)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0275906
|
Thermo Scientific Chemicals
L0275906 |
5 g |
Each for $89.11
|
|
|||||
AAL0275914
|
Thermo Scientific Chemicals
L0275914 |
25 g |
Each for $302.44
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
33499-42-2 | |
178.231 | |
RJYPOWRKMKNFHH-UHFFFAOYSA-N | |
118492 | |
CCOC(=O)C1=C(C=C(C=C1)C)C |
C11H14O2 | |
MFCD00015438 | |
2,4-dimethylbenzoic acid ethyl ester, benzoic acid, 2,4-dimethyl-, ethyl ester, acmc-20al1j, 2,4-dimethyl-benzoic acid ethyl ester, benzoic acid,2,4-dimethyl-, ethyl ester | |
ethyl 2,4-dimethylbenzoate |
Specifications
33499-42-2 | |
1.513 | |
MFCD00015438 | |
2,4-dimethylbenzoic acid ethyl ester, benzoic acid, 2,4-dimethyl-, ethyl ester, acmc-20al1j, 2,4-dimethyl-benzoic acid ethyl ester, benzoic acid,2,4-dimethyl-, ethyl ester | |
RJYPOWRKMKNFHH-UHFFFAOYSA-N | |
ethyl 2,4-dimethylbenzoate | |
118492 | |
≥98% |
5 g | |
C11H14O2 | |
1938737 | |
Not miscible or difficult to mix with water. | |
CCOC(=O)C1=C(C=C(C=C1)C)C | |
178.231 | |
178.23 | |
Ethyl 2,4-dimethylbenzoate |
Safety and Handling
EINECSNumber : 251-544-2
TSCA : No
Recommended Storage : Ambient temperatures