missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2,4-Dihydroxy-6-methylnicotinate, TCI America™

Manufacturer:  TCI America E08695G

 View more versions of this product

Catalog No. E08695G



Specifications

Specifications

Ethyl 2,4-Dihydroxy-6-methylnicotinate
210°C
70254-52-3
MFCD02929397
CMCZAWPDHHYFPU-UHFFFAOYSA-N
ethyl 4-hydroxy-6-methyl-2-oxo-1H-pyridine-3-carboxylate
54676534
Crystal-Powder at 20°C
White
5g
C9H11NO4
ethyl 2,4-dihydroxy-6-methylnicotinate, ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate, ethyl 2,4-dihydroxy-6-methylpyridine-3-carboxylate, 2,4-dihydroxy-6-methylnicotinic acid ethyl ester, ethyl2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 2,4-dihydroxy-6-methyl-3-pyridine-carboxylate, 2,4-dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, 4-hydroxy-6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid ethyl ester, 2,4-dihydroxy-6-methyl-nicotinic acid ethyl ester
CCOC(=O)C1=C(C=C(NC1=O)C)O
197.19
197.19
≥98.0% (GC,T)

Chemical Identifiers

70254-52-3
197.19
CMCZAWPDHHYFPU-UHFFFAOYSA-N
54676534
CCOC(=O)C1=C(C=C(NC1=O)C)O
C9H11NO4
MFCD02929397
ethyl 2,4-dihydroxy-6-methylnicotinate, ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate, ethyl 2,4-dihydroxy-6-methylpyridine-3-carboxylate, 2,4-dihydroxy-6-methylnicotinic acid ethyl ester, ethyl2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 2,4-dihydroxy-6-methyl-3-pyridine-carboxylate, 2,4-dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, 4-hydroxy-6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid ethyl ester, 2,4-dihydroxy-6-methyl-nicotinic acid ethyl ester
ethyl 4-hydroxy-6-methyl-2-oxo-1H-pyridine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents