Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4-dihydroxy-6-methylnicotinate, 98%

$157.74 - $157.74

Chemical Identifiers

Chemical Name or Material Ethyl 2,4-dihydroxy-6-methylnicotinate
CAS 70254-52-3
Molecular Formula C9H11NO4
Formula Weight 197.20g/mol
MDL Number MFCD02929397
Synonym 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester; Ethyl 2,4-dihydroxy-6-methylpyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2714806

 
alfa aesar™
H27148-06
5g Each for $157.74
Description & Specifications

Specifications

Melting Point 212° to 213°C
Quantity 5g
Chemical Name or Material Ethyl 2,4-dihydroxy-6-methylnicotinate
CAS 70254-52-3
Assay 98%
Molecular Formula C9H11NO4
Formula Weight 197.20g/mol
MDL Number MFCD02929397
Synonym 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester; Ethyl 2,4-dihydroxy-6-methylpyridine-3-carboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.