missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2,4-Difluorobenzoate, TCI America™

Manufacturer:  TCI America E093525G

 View more versions of this product

Catalog No. E093525G



Specifications

Specifications

Ethyl 2,4-Difluorobenzoate
Yellow
108928-00-3
MFCD00153149
OPQFYGPAOVCNEQ-UHFFFAOYSA-N
ethyl 2,4-difluorobenzoate
2737170
Clear Liquid at 20°C
88°C
25g
C9H8F2O2
2,4-difluorobenzoic acid ethyl ester, 2,4-difluoro-benzoic acid ethyl ester, pubchem3467, acmc-209w2m, ethyl-2,4-difluorobenzoate, ethyl 2,4-di-fluorobenzoate, rarechem al bi 0213, ethyl2,4-difluorobenzoate, 2,4 ethyl-difluorobenzoate, 2,4-difluorobenzoic acid, ethyl ester
CCOC(=O)C1=C(C=C(C=C1)F)F
186.158
186.16
≥98.0% (GC)

Chemical Identifiers

108928-00-3
186.158
OPQFYGPAOVCNEQ-UHFFFAOYSA-N
2737170
CCOC(=O)C1=C(C=C(C=C1)F)F
C9H8F2O2
MFCD00153149
2,4-difluorobenzoic acid ethyl ester, 2,4-difluoro-benzoic acid ethyl ester, pubchem3467, acmc-209w2m, ethyl-2,4-difluorobenzoate, ethyl 2,4-di-fluorobenzoate, rarechem al bi 0213, ethyl2,4-difluorobenzoate, 2,4 ethyl-difluorobenzoate, 2,4-difluorobenzoic acid, ethyl ester
ethyl 2,4-difluorobenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents