Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4-difluorobenzoate, 98%

$168.76 - $168.76

Chemical Identifiers

Chemical Name or Material Ethyl 2,4-difluorobenzoate
CAS 108928-00-3
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD00153149
Synonym 2,4-Difluorobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1317706

  SDS
alfa aesar™
A1317706
5g Each for $168.76
Description & Specifications

Specifications

Formula Weight 186.16
Chemical Name or Material Ethyl 2,4-difluorobenzoate
Boiling Point 65° to 67°C (15mmHg)
Assay 98%
Molecular Formula C9H8F2O2
MDL Number MFCD00153149
CAS 108928-00-3
Synonym 2,4-Difluorobenzoic acid ethyl ester
Quantity 5g