Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,4,6-triisopropylbenzoate, 99%

$26.23 - $313.41

Chemical Identifiers

Chemical Name or Material Ethyl 2,4,6-triisopropylbenzoate
CAS 63846-76-4
Molecular Formula C18H28O2
Formula Weight 276.42
MDL Number MFCD03788745
Beilstein 278852
Synonym 2,4,6-Triisopropylbenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1966603

  SDS
alfa aesar™
L1966603
1g Each for $26.23

AAL1966606

  SDS
alfa aesar™
L1966606
5g Each for $81.81

AAL1966614

  SDS
alfa aesar™
L1966614
25g Each for $313.41
Description & Specifications

Specifications

Formula Weight 276.42
Chemical Name or Material Ethyl 2,4,6-triisopropylbenzoate
MDL Number MFCD03788745
Assay 99%
Molecular Formula C18H28O2
Beilstein 278852
Quantity 1g
Synonym 2,4,6-Triisopropylbenzoic acid ethyl ester
CAS 63846-76-4