missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(3-pyridyl)thiazole-4-acetate, 97%

Manufacturer:  Alfa Aesar™ H54753MD

 View more versions of this product

Catalog No. AAH54753MD



Specifications

Specifications

Ethyl 2-(3-pyridyl)thiazole-4-acetate
80653-69-6
MFCD06739823
2-3-pyridyl thiazole-4-acetic acid ethyl ester
CCOC(=O)CC1=CSC(=N1)C2=CN=CC=C2
248.3
248.3
250mg
C12H12N2O2S
991826
JGRCACXOIKCVIX-UHFFFAOYSA-N
ethyl 2-(2-pyridin-3-yl-1,3-thiazol-4-yl)acetate
7176349
97%

Chemical Identifiers

80653-69-6
248.3
JGRCACXOIKCVIX-UHFFFAOYSA-N
7176349
CCOC(=O)CC1=CSC(=N1)C2=CN=CC=C2
C12H12N2O2S
MFCD06739823
2-3-pyridyl thiazole-4-acetic acid ethyl ester
ethyl 2-(2-pyridin-3-yl-1,3-thiazol-4-yl)acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents