Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(3-N-phthalimidopropyl)acetoacetate, 95%

$253.99 - $253.99

Chemical Identifiers

Chemical Name or Material Ethyl 2-(3-N-phthalimidopropyl)acetoacetate
CAS 55747-45-0
Molecular Formula C17H19NO5
Formula Weight 317.34
MDL Number MFCD00399474
Synonym Ethyl acetopropylphthalimide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1472414

 
alfa aesar™
L1472414
25g Each for $253.99
Description & Specifications

Specifications

Formula Weight 317.34
Chemical Name or Material Ethyl 2-(3-N-phthalimidopropyl)acetoacetate
Assay 95%
Molecular Formula C17H19NO5
MDL Number MFCD00399474
CAS 55747-45-0
Synonym Ethyl acetopropylphthalimide
Melting Point 62° to 64°C
Quantity 25g