missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(3-hydroxyphenyl)-4-methylthiazole-5-carboxylate, 97%

$74.41 - $982.00

Chemical Identifiers

CAS 209538-87-4
Molecular Formula C13H13NO3S
Molecular Weight (g/mol) 263.311
MDL Number MFCD20265360
InChI Key RBLKTMVSJHHLGF-UHFFFAOYSA-N
Synonym 2-3-hydroxyphenyl-4-methylthiazole-5-carboxylic acid ethyl ester
PubChem CID 57495753
IUPAC Name ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CC(=CC=C2)O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54868MD View Documents Alfa Aesar™
H54868MD
250mg Each for $74.41
 
AAH5486803 View Documents Alfa Aesar™
H5486803
1g Each for $236.26
 
AAH5486806 View Documents Alfa Aesar™
H5486806
5g Each for $982.00
 
Specifications

Chemical Identifiers

209538-87-4
263.311
RBLKTMVSJHHLGF-UHFFFAOYSA-N
57495753
CCOC(=O)C1=C(N=C(S1)C2=CC(=CC=C2)O)C
C13H13NO3S
MFCD20265360
2-3-hydroxyphenyl-4-methylthiazole-5-carboxylic acid ethyl ester
ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate

Specifications

154°C to 158°C
209538-87-4
MFCD20265360
RBLKTMVSJHHLGF-UHFFFAOYSA-N
ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
57495753
97%
250mg
C13H13NO3S
2-3-hydroxyphenyl-4-methylthiazole-5-carboxylic acid ethyl ester
CCOC(=O)C1=C(N=C(S1)C2=CC(=CC=C2)O)C
263.311
263.31
Ethyl 2-(3-hydroxyphenyl)-4-methylthiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen