missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(3-hydroxyphenyl)-4-methylthiazole-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H54868MD

 View more versions of this product

Catalog No. AAH54868MD



Specifications

Specifications

Ethyl 2-(3-hydroxyphenyl)-4-methylthiazole-5-carboxylate
250mg
C13H13NO3S
2-3-hydroxyphenyl-4-methylthiazole-5-carboxylic acid ethyl ester
CCOC(=O)C1=C(N=C(S1)C2=CC(=CC=C2)O)C
263.311
263.31
154°C to 158°C
209538-87-4
MFCD20265360
RBLKTMVSJHHLGF-UHFFFAOYSA-N
ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
57495753
97%

Chemical Identifiers

209538-87-4
263.311
RBLKTMVSJHHLGF-UHFFFAOYSA-N
57495753
CCOC(=O)C1=C(N=C(S1)C2=CC(=CC=C2)O)C
C13H13NO3S
MFCD20265360
2-3-hydroxyphenyl-4-methylthiazole-5-carboxylic acid ethyl ester
ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents