Please login to your online account to display your discounted pricing

Ethyl 2,3-dibromopropionate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2, 3-dibromopropionate
Name Note 99%
CAS 3674-13-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H8Br2O2
Linear Formula BrCH2CH(Br)CO2C2H5
Formula Weight 259.91
MDL Number MFCD00000212
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118370250

  SDS
thermo fisher scientific
118370250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 259.91
Quantity 25mL
Color Clear colorless to yellow
Density 1.7880g/mL
EINECS Number 222-941-8
TSCA TSCA
RTECS Number UA2458382
Beilstein 02, 259
Chemical Name or Material Ethyl 2, 3-dibromopropionate
Packaging Glass bottle
Refractive Index 1.4976 to 1.5016 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C5H8Br2O2
Linear Formula BrCH2CH(Br)CO2C2H5
MDL Number MFCD00000212
CAS 3674-13-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic