Please login to your online account to display your discounted pricing

Ethyl 2-(3,4-dichlorophenyl)-2-oxoacetate, Technical, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 2-(3, 4-dichlorophenyl)-2-oxoacetate
CAS 34966-52-4
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C10H8Cl2O3
Formula Weight 247.08
MDL Number MFCD01934864
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB07805DA

  SDS
fisher scientific company
SPB07805DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.08
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
CAS 34966-52-4
Molecular Formula C10H8Cl2O3
Chemical Name or Material Ethyl 2-(3, 4-dichlorophenyl)-2-oxoacetate
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Amber glass bottle
MDL Number MFCD01934864
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.