Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,3,4,5-tetrafluorobenzoate, 98%

$64.19 - $227.81

Chemical Identifiers

Chemical Name or Material Ethyl 2,3,4,5-tetrafluorobenzoate
CAS 122894-73-9
Molecular Formula C9H6F4O2
Formula Weight 222.14
MDL Number MFCD02094158
Synonym 2,3,4,5-Tetrafluorobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2192004

 
alfa aesar™
B2192004
2g Each for $64.19

AAB2192009

 
alfa aesar™
B2192009
10g Each for $227.81
Description & Specifications

Specifications

Formula Weight 222.14
Chemical Name or Material Ethyl 2,3,4,5-tetrafluorobenzoate
Assay 98%
Molecular Formula C9H6F4O2
MDL Number MFCD02094158
CAS 122894-73-9
Synonym 2,3,4,5-Tetrafluorobenzoic acid ethyl ester
Quantity 2g