Learn More
Ethyl 2,2-difluoropropionate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3185206
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications
Ethyl 2,2-difluoropropionate | |
1.102 | |
C5H8F2O2 | |
MFCD06657970 | |
UN3272 | |
Insoluble in water. | |
CCOC(=O)C(C)(F)F | |
138.114 | |
138.11 |
28781-85-3 | |
24°C (20 mmHg) | |
CH3CF2CO2CH2CH3 | |
5 g | |
2,2-difluoropropionic acid ethyl ester, ethyl 2,2-difluoropropionate, 2,2-difluoropropionicacidethylester, 2,2-difluoro-propionic acid ethyl ester, propanoic acid, 2,2-difluoro-, ethyl ester, ethyl difluoropropionate, ethyl 2,2 difluoropropionate, difluoropropionic acid ethyl ester, 2,2-difluoropropionic acidethyl ester | |
ISVLDAIKRGXNCZ-UHFFFAOYSA-N | |
ethyl 2,2-difluoropropanoate | |
2761188 | |
97% |
Chemical Identifiers
28781-85-3 | |
138.114 | |
ISVLDAIKRGXNCZ-UHFFFAOYSA-N | |
2761188 | |
CCOC(=O)C(C)(F)F |
C5H8F2O2 | |
MFCD06657970 | |
2,2-difluoropropionic acid ethyl ester, ethyl 2,2-difluoropropionate, 2,2-difluoropropionicacidethylester, 2,2-difluoro-propionic acid ethyl ester, propanoic acid, 2,2-difluoro-, ethyl ester, ethyl difluoropropionate, ethyl 2,2 difluoropropionate, difluoropropionic acid ethyl ester, 2,2-difluoropropionic acidethyl ester | |
ethyl 2,2-difluoropropanoate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only