missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2,2-diethylbutyrate, 98%

$94.24 - $332.09

Chemical Identifiers

CAS 34666-17-6
Molecular Formula C10H20O2
Formula Weight 172.27
MDL Number MFCD00154424
Beilstein 1757834
Synonym 2,2-Diethylbutyric acid ethyl ester; Ethyl triethylacetate
Chemical Name or Material Ethyl 2,2-diethylbutyrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1997203 SDS Alfa Aesar™
L1997203
1g Each for $94.24
AAL1997206 SDS Alfa Aesar™
L1997206
5g Each for $332.09
Specifications

Description

Applications
Ethyl 2,2-diethylbutyrate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

34666-17-6
172.27
1757834
Ethyl 2,2-diethylbutyrate
C10H20O2
MFCD00154424
2,2-Diethylbutyric acid ethyl ester; Ethyl triethylacetate

Specifications

1g
85° to 87°C (30mmHg)
172.27
1757834
Ethyl 2,2-diethylbutyrate
34666-17-6
C10H20O2
MFCD00154424
2,2-Diethylbutyric acid ethyl ester; Ethyl triethylacetate
98%