Please login to your online account to display your discounted pricing

Ethyl 2-(2-aminothiazol-4-yl)-2-hydroxyiminoacetate, 97%, pure, predominantly syn, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-(2-aminothiazol-4-yl)-2-hydroxyiminoacetate
Name Note 97%
CAS 64485-82-1
Purity Grade Extra Pure
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H9N3O3S
Formula Weight 215.2
MDL Number MFCD00010415
Synonym Ethyl 2-amino-α-(hydroxyimino)-4-thiazoleacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC247891000

  SDS
thermo fisher scientific
247891000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 215.2
Purity Grade Extra Pure
Physical Form Fine Powder
Quantity 100g
Color Light yellow to beige
Melting Point 187° to 192°C
EINECS Number 264-910-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 1 MOISTURE SENSITIVE
Synonym Ethyl 2-amino-α-(hydroxyimino)-4-thiazoleacetate
Assay Percent Range 97%
Molecular Formula C7H9N3O3S
Name Note 97%
MDL Number MFCD00010415
CAS 64485-82-1
Chemical Name or Material Ethyl 2-(2-aminothiazol-4-yl)-2-hydroxyiminoacetate
Safety and Handling

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.