Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-(2-amino-5-nitrophenyl)-4-methylthiazole-5-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-(2-amino-5-nitrophenyl)-4-methylthiazole-5-carboxylate
CAS 41450-33-3
Formula Weight 307.33
Synonym 2-(2-Amino-5-nitrophenyl)-4-methylthiazole-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6699303

 
alfa aesar™
H6699303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66993MD

 
alfa aesar™
H66993MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.33
Melting Point 238° to 242°C
Chemical Name or Material Ethyl 2-(2-amino-5-nitrophenyl)-4-methylthiazole-5-carboxylate
CAS 41450-33-3
Physical Form Crystalline Solid
Assay 97%
Synonym 2-(2-Amino-5-nitrophenyl)-4-methylthiazole-5-carboxylic acid ethyl ester
Color Yellow
Quantity 1g