missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate, TCI America™

Manufacturer:  TCI America E115925G

 View more versions of this product

Catalog No. E115925G



Specifications

Specifications

Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate
146°C
64987-08-2
MFCD00010414
XNVRKLCQBZTGNA-UHFFFAOYSA-N
ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-oxoacetate
4356513
Crystal-Powder at 20°C
Yellow
25g
C7H8N2O3S
2-(2-Amino-4-thiazolyl)-2-oxoacetic Acid Ethyl Ester
CCOC(=O)C(=O)C1=CSC(=N1)N
200.212
200.21
≥96.0% (HPLC,T)

Chemical Identifiers

64987-08-2
200.212
XNVRKLCQBZTGNA-UHFFFAOYSA-N
4356513
CCOC(=O)C(=O)C1=CSC(=N1)N
C7H8N2O3S
MFCD00010414
2-(2-Amino-4-thiazolyl)-2-oxoacetic Acid Ethyl Ester
ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-oxoacetate
Safety and Handling

Safety and Handling

TSCA : No

Documents