missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1H-Tetrazole-5-acetate, TCI America™

Manufacturer:  TCI America E08595G

 View more versions of this product

Catalog No. E08595G



Specifications

Specifications

Ethyl 1H-Tetrazole-5-acetate
128°C
13616-37-0
MFCD00274213
NAOHMNNTUFFTBF-UHFFFAOYSA-N
ethyl 2-(2H-tetrazol-5-yl)acetate
543616
Crystal-Powder at 20°C
White
5g
C5H8N4O2
1H-Tetrazole-5-acetic Acid Ethyl Ester
CCOC(=O)CC1=NNN=N1
156.145
156.15
≥98.0% (HPLC,T)

Chemical Identifiers

13616-37-0
156.145
NAOHMNNTUFFTBF-UHFFFAOYSA-N
543616
CCOC(=O)CC1=NNN=N1
C5H8N4O2
MFCD00274213
1H-Tetrazole-5-acetic Acid Ethyl Ester
ethyl 2-(2H-tetrazol-5-yl)acetate
Safety and Handling

Safety and Handling

TSCA : No

Documents