Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1-piperazineacetate, 96%

Chemical Identifiers

Chemical Name or Material Ethyl 1-piperazineacetate
CAS 40004-08-8
Molecular Formula C8H16N2O2
Formula Weight 172.23
MDL Number MFCD00040739
UN Number UN2735
Beilstein 743260
Synonym N-(Carboethoxymethyl)piperazine; 1-(Ethoxycarbonylmethyl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0731214

 
alfa aesar™
L0731214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0731206

 
alfa aesar™
L0731206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.23
Chemical Name or Material Ethyl 1-piperazineacetate
Assay 96%
CAS 40004-08-8
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H16N2O2
MDL Number MFCD00040739
Beilstein 743260
Synonym N-(Carboethoxymethyl)piperazine; 1-(Ethoxycarbonylmethyl)piperazine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN2735
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Boiling Point 127°C (13mmHg)
Quantity 25g

Applications
Ethyl 1-piperazineacetate is used as a pharmaceutical intermediate.

Solubility
Soluble in water, chloroform, and methanol.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.