missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-methylnipecotate, +98%, ACROS Organics™

$255.65 - $255.65

Chemical Identifiers

CAS 5166-67-6
Assay Percent Range 98+%
Molecular Formula C9H17NO2
Formula Weight 171.24
MDL Number MFCD00006496
Synonym Ethyl 1-methyl-3-piperidinecarboxylate
Chemical Name or Material Ethyl 1-methylnipecotate
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC186610250 SDS Thermo Fisher Scientific
AC186610250
Glass bottle 25g Each for $255.65
Specifications Description & Specifications

Chemical Identifiers

5166-67-6
C9H17NO2
MFCD00006496
Ethyl 1-methylnipecotate
98+%
171.24
Ethyl 1-methyl-3-piperidinecarboxylate
≥98.0%

Specifications

Clear colorless to pale yellow
5166-67-6
Authentic
1.4490 to 1.4510 (20°C, 589nm)
98+%
171.24
22, V,1, 237
Ethyl 1-methylnipecotate
25g
0.9540g/mL
Glass bottle
Liquid
C9H17NO2
MFCD00006496
Ethyl 1-methyl-3-piperidinecarboxylate
≥98.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.