missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 1-methylimidazole-2-carboxylate, 95%

Manufacturer:  Alfa Aesar™ H6118506

 View more versions of this product

Catalog No. AAH6118506



Description

Description

Applications
Ethyl 1-methylimidazole-2-carboxylate is used as a synthetic intermediate for solid phase synthesis of polyamides containing imidazole.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 1-methylimidazole-2-carboxylate
30148-21-1
MFCD02683326
Soluble in dichloromethane,ether,ethyl acetate and methanol.
CCOC(=O)C1=NC=CN1C
154.169
154.17
95%
5g
C7H10N2O2
1-methyl-1h-imidazole-2-carboxylic acid ethyl ester
NOTZYDYZBOBDFE-UHFFFAOYSA-N
ethyl 1-methylimidazole-2-carboxylate
4261883
Solid

Chemical Identifiers

30148-21-1
154.169
NOTZYDYZBOBDFE-UHFFFAOYSA-N
4261883
CCOC(=O)C1=NC=CN1C
C7H10N2O2
MFCD02683326
1-methyl-1h-imidazole-2-carboxylic acid ethyl ester
ethyl 1-methylimidazole-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents